Learn More
S Murray and Co™ Heavy Duty Scissors
Use these general purpose scissors for cutting anything from bandages to stainless steel mesh. S Murray™ Heavy Duty Scissors are heavy duty but lightweight with stainless steel blades and robust construction.
Brand:  S Murray and Co™ E916/05
Product Code. 12952065
  • 31.25 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Nickel plated steel
  • Straight with one sharp point
  • For cutting anything from bandages to stainless steel mesh
  • Autoclavable
Specifications
Specifications
Heavy Duty Scissors
180 mm
Autoclavable
Straight
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
S Murray and Co™ Heavy Duty Scissors > Length: 180mm