missing translation for 'onlineSavingsMsg'
Learn More

Manutan Heavy duty shelving, Rapid 1, blue/orange, 1830mm x 1830mm x 455mm

Brand:  Manutan A361054

Encompass_Preferred

Product Code. 12891879

  • 457.38 € / Each
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Strong, sturdy shelving unit with 400kg capacity per shelf. Ideal for large, bulky loads. Quick, easy boltless assembly - frames and beams simply clip together and only require a rubber mallet. High density chipboard shelves with max shelf capacity of 400kg. Manufactured using European structural quality steel. Hardwearing powder coated finish. Load capacity TUV certified - test to establish safe working loads. Supplied flat-packed.
  • Number of shelves : 4
  • Shelf capacity UDL: 400kg
  • Dimensions (H x W x D): 1830mm x 1830mm x 455mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.