Learn More
Heidolph™ Hei-FLOW Core 120
Peristaltic pump ideal for the simple conveying of all media.
Brand:  Heidolph™ 523-50010-02
Product Code. 17363000
  • 2980.00 € / Each
Explore more special offers
Description
Description
  • The pump drive of this model makes it possible to switch between a single channel and multichannel pump head. By simply flipping the switch, it can also be conveyed in an anti-clockwise direction.
  • Depending on the adapted pump head, a flow rate of 0.83 to 861 mL/min for the single-channel and 0.005 to 364 mL/min for the multichannel pump head is guaranteed. The Value 01 conveys with a control accuracy of ± 5% at a speed range of 10 to 120 rpm.
Specifications
Specifications
UK Plug
Analog
230 V
Digital Variable Speed
IP55