Learn More
Heidolph™ Hei-FLOW Core 600
This pump of the Hei-Flow Value series is particularly suitable when high volumes need to be transported easily.
Brand:  Heidolph™ 523-50060-02
Product Code. 17393000
  • 2770.00 € / Each
Explore more special offers
Description
Description
  • This pump of the Hei-Flow Value series is particularly suitable when high volumes need to be transported easily. A flow rate of 4.0 to 4151 mL/min can be set for this purpose. This model is designed for the use of single-channel pump heads.
  • Even with a load change, the speed of 50 to 600 rpm is kept constant at a control accuracy of ±5%. It is easy to switch to the reverse direction of rotation.
Specifications
Specifications
UK Plug
Analog
230 V
Digital Variable Speed
IP55