Learn More
Heidolph™ Hei-FLOW Ultimate 120 Multi
Model that allows you to choose between a single-channel or multi-channel pump head.
Brand:  Heidolph™ 523-52013-02
Product Code. 17363010
  • 4095.00 € / Each
Explore more special offers
Description
Description
  • Accurate dosing is particularly important in the pharmaceutical, cosmetic or food sector. Comprehensible process conditions are a prerequisite for the safe conveyance of sensitive media. This is ensured by a control accuracy of ±1% and precise speed adjustment (5 to 120 rpm).
  • In addition, an optional foot switch allows the pump to be operated in closed exhaust hoods. The parameters - speed, direction of rotation and on/off function - can be controlled externally either through the analog or digital interface.
  • In this model, you can choose between a single-channel or multichannel pump head. Flow rates vary depending on the pump head: the single-channel head has a flow rate of 0.38 to 813 mL/min; for the multichannel head 0.005 to 364 mL/min. Depending on the focus of the process, the following parameters can be set on the device: Speed, tubing diameter, flow rate display, metering volume, interval dosing and break times. The pump can be calibrated for optimum dosing results.
Specifications
Specifications
UK Plug
Analog
Variable-Speed Digital
230 V
Digital Variable Speed
IP55
IP 55
Analog