missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Hei-TORQUE Ultimate 200

The Hei-TORQUE Precision line are Heidolph high performance overhead stirrers.

Brand:  Heidolph™ 501-62020-03

2074.88 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.


Product Code. 17342990

  • 2560.00 € / Each
Estimated Shipment: 07-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The Hei-TORQUE Precision 200 laboratory stirrer is ideal for the powerful stirring of viscosities up to 100,000 mPas or volumes up to 50 liters (H2O). It has a wide speed range (10 to 2000 rpm) and a maximum torque of 200 Ncm.
  • In addition, the Hei-TORQUE Precision 200 offers, above all, the possibility of ramp programming, process monitoring (including timer function, torque trend display as a value), as well as the necessary equipment for documenting your application parameters. For this it is equipped with two interfaces - USB and RS232 - which give you the greatest flexibility to connect the device to a PC, a programmable logic controller (PLC) or an automation system such as e.g. LabManager. The scope of delivery includes the free Hei-Control software for remote control of up to 4 devices via PC.
  • The laboratory stirrer Hei-TORQUE Value 200 performs every stirring task with minimal noise thanks to the latest engine technology. The safety-related start/stop touch-function serves to minimize the risk of unintentional switching on.
  • Suitable for use in fume hoods or as a drive for reactor super structures, the Hei-TORQUE Precision 200 has a quick -action chuck (quick chuck with safety ring) with a diameter of 10.5 mm, which can be combined with a large number of stirring tools for the best stirring results.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

230 V
UK
200 Ncm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heidolph™ Hei-TORQUE Ultimate 200 > UK Plug

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.