Learn More
Heidolph™ Instruments Hei-VAP Core All-round Chill Package, UK-plug
The Hei-VAP University Academic Package features a hand lift, vacuum pump, chiller, and tubing set.
Brand: Heidolph™ Instruments 571-21300-03
This item is not returnable.
View return policy
Description
- Hei-VAP Core with hand lift and vertical glassware set G3
- Hei-VAC Control
- Vacuum pump Rotavac Valve Tec
- Tubing set
- Chiller Hei-CHILL 250
- Thermal fluid Kyro 30 (10 L)
Specifications
Hand Lift Rotary Evaporator | |
UK-plug |