Learn More
Heidolph™ Instruments Hei-VAP Powder Drying Package, UK-plug
The Hei-VAP Powder Drying Package features a motor lift, powder flask, and tubing set.
Brand:  Heidolph™ Instruments 574-32300-03
Product Code. 17363110
  • 9830.00 € / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Hei-VAP Expert Control with motor lift and vertical glassware set G3
  • Vacuum pump Rotavac Valve Control
  • Vacuum valve
  • Tubing set
  • Powder flask 2 L
  • Vapor tube with powder filter
Specifications
Specifications
Motor Lift Rotary Evaporator
UK-plug