Learn More
Heidolph™ Instruments Hei-VAP Ultimate ML G5, UK-plug
The Hei-VAP Ultimate ML G5 allows for precise setting of rotation speed and heating bath temperature on the digital touch display or directly via the knobs on the operating panel.
Brand: Heidolph™ Instruments 576-01500-03
This item is not returnable.
View return policy
Description
Precise setting of rotation speed and heating bath temperature on the digital touch display or – for quick access during an active process – directly via the knobs on the operating panel.- Central operating panel with 7 in. touch display
- Motor lift model with G5 dry ice condenser
- Clear display of all values (Basic and Advanced modes)
- With timer, limitation of max. temperature, multilingual menu and text display for error messages
- Separate knobs for rotation and heating with locking function to prevent accidental readjusting
- LED ring lights and pictogram to display the activity even from a distance
- Integrated solvent library to facilitate adjustment of the required vacuum on an external vacuum controller
- USB and Micro SD plus LAN / RS 232 interfaces (Lab 4.0 ready)
- Optional display of vapor temperature
- Subsequent upgrade to Hei-VAP Ultimate Control possible, including vacuum and chiller control
Specifications
5 L | |
Motor Lift Rotary Evaporator | |
UK-plug |