missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Height Adjustable Floor Stand for Herasafe™ 2030i

For use with Herasafe 2030i Biological Safety Cabinets

Brand:  Thermo Scientific™ 50155688

1050.23 EUR valid until 2025-12-31
Use promo code "25057" to get your promotional price.


 View more versions of this product

Product Code. 15983110

  • 1105.50 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Height Adjustable Floor Stand in increments of 50mm.(Height range 680-880mm), charcoal grey

  • Adjustment steps of 50mm
  • Charcoal grey
  • Height - 680-880mm
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Adjustable Floor Stand (Height of 680-880mm), 0.9 m, charcoal grey
26.7 in. to 34.6 in.
1000 mm
800 mm
Height Adjustable Floor Stand
680 to 880 mm
Herasafe 2030i Biological Safety Cabinet size 0.9 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.