Learn More
Helios Italquartz Quartz Wool
For filtration and insulation in environments where high quartz purity properties are needed. Helios Italquartz™ Quartz Wool is chemically inert.
Brand:  Helios Italquartz 39P00505
Product Code. 11755694
  • 98.50 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Supplied in 50g amounts
  • High temperature resistance
  • Resistant to corrosion
Specifications
Specifications
50g
Quartz wool fiber
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Helios Italquartz Quartz Wool > Weight: 50g