missing translation for 'onlineSavingsMsg'
Learn More

Hellma™ Hellmanex III Concentrated Liquid Cleaning Solution

Product Code. 12701725
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for the effective cleaning of glass or silica cells and other sensitive optical components. Hellma™ Hellmanex III Concentrated Liquid Cleaning Solution is an alkaline liquid.

  • Reduces surface tension of water
  • Good wetting action
  • High emulsifying and dispersing capabilities
  • With dosing feeder

Specifications

Product Type Cleaning Solution
For Use With (Application) Optical components
Volume (Metric) 1 L
compliance-icons compliance-icons
Product Identifier
  • Hellmanex lll
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : TRIPOTASSIUM ORTHOPHOSPHATE
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'