Learn More
Hirschmann™ Hematocrit Tubes, NH4-heparinized
Excellent for centrifuging blood in the laboratory. Hirschmann™ Hematocrit Tubes are manufactured from disposable capillary glass.
Brand:  Hirschmann™ 9100418
Product Code. 30090385
  • 405.00 € / Pack of 5000
Explore more special offers
Description
Description
  • Disposable
  • With ringmark at 60mm
  • NH4-heparinized
  • Color-code green
  • CE acc. IVD 98/79 EC
Specifications
Specifications
Hematocrit Tube NH4-hep
0.5/0.6 mm
75 mm
18 μL
For centrifuging blood samples, Ammonium heparinized
CE, IVD 98/79 EC