missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Sodium Heparinized Hematocrit Tubes
Excellent for centrifuging blood in the laboratory. Hirschmann Instruments™ Sodium Heparinized Hematocrit Tubes are manufactured from disposable capillary glass.
29.45 € - 276.00 €
Specifications
Product Type | Hematocrit Tube Na-hep |
---|---|
For Use With (Application) | For centrifuging blood samples, Sodium heparinized |
Certifications/Compliance | DIN EN ISO 12772, CE, IVD 98/79 EC |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10233391
|
Hirschmann™
9100299 |
100 μL | N/A | ||||||
30090390
|
Hirschmann™
9100299 |
100 μL |
276.00 €
Pack of 5000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10410772
|
Hirschmann™
9100260 |
60 μL |
29.45 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30090388
|
Hirschmann™
9100260 |
60 μL |
151.00 €
Pack of 5000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10199641
|
Hirschmann™
9100275 |
75 μL |
29.45 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30090387
|
Hirschmann™
9100275 |
75 μL |
152.00 €
Pack of 5000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Color-coded red
- Acc. to DIN EN ISO 12772, CE acc. IVD 98/79 EC
Specifications
Hematocrit Tube Na-hep | |
For centrifuging blood samples, Sodium heparinized | |
DIN EN ISO 12772, CE, IVD 98/79 EC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title