missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Hematoxylin
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
230.00 € - 549.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30018325
|
Melford
H12000-25.0 |
25 g |
230.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30018324
|
Melford
H12000-100.0 |
100 g |
549.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 517-28-2
- Molecular Formula: C16H14O6 3H2O
- Molecular Weight: 356.3
Specifications
517-28-2 | |
C16H14O6-3H2O |