missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Hematoxylin, 85%
123.62 € - 1240.17 €
Chemical Identifiers
CAS | 517-28-2 |
---|---|
Molecular Formula | C16H14O6 |
Molecular Weight (g/mol) | 302.28 |
MDL Number | MFCD00078111 |
InChI Key | WZUVPPKBWHMQCE-WKTCHCBJNA-N |
Synonym | Natural Black 1, C.I. 75290 |
PubChem CID | 45029742 |
IUPAC Name | (6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol |
SMILES | [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10548512
|
Thermo Scientific Chemicals
411160250 |
25 g | Glass Bottle |
123.62 €
25g |
In Stock
Log in to see stock. |
||||
10239600
|
Thermo Scientific Chemicals
411161000 |
100 g | Glass bottle |
350.75 €
100g |
In Stock
Log in to see stock. |
||||
11966921
|
Thermo Scientific Chemicals
411165000 |
500 g | Glass bottle |
1240.17 €
500g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
517-28-2 | |
302.28 | |
WZUVPPKBWHMQCE-WKTCHCBJNA-N | |
45029742 | |
[H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3 |
C16H14O6 | |
MFCD00078111 | |
Natural Black 1, C.I. 75290 | |
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol |
Specifications
100.0°C to 120.0°C | |
85% | |
MFCD00078111 | |
Solubility in water: slightly soluble; soluble in hot water;. Other solubilities: soluble in hot alcohol, glycerol, borax, alkali hy, droxides (aq. solution purple red; becomes blue vi, olett during storage at air). | |
Authentic | |
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol | |
45029742 | |
10% (105°C) max. | |
Pure | |
Beige to Brown | |
Hematoxylin |
517-28-2 | |
C16H14O6 | |
Natural Black 1, C.I. 75290 | |
WZUVPPKBWHMQCE-WKTCHCBJNA-N | |
[H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3 | |
302.28 | |
302.27 | |
85% | |
88% (on dry substance) min. | |
Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Hematoxylin
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Hematoxylin
RUO – Research Use Only