missing translation for 'onlineSavingsMsg'
Learn More
Hemin (porcine), 97+%, Thermo Scientific Chemicals Product Code.: 11499093

Hemin (porcine), 97+%, Thermo Scientific Chemicals

Product Code. 11499093
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11499093

Brand: Thermo Scientific Alfa Aesar A11165.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hemin is used in cellular protection and control mechanism. It stimulates the synthesis of globulin. It is also used to study the bacterial strains and growth conditions, in the short-term colony assays and cell transfections. Further, it is used to treat the symptoms of various porphyrias. In addition, it is used in the preparation of agar medium for the bacteria culture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hemin is used in cellular protection and control mechanism. It stimulates the synthesis of globulin. It is also used to study the bacterial strains and growth conditions, in the short-term colony assays and cell transfections. Further, it is used to treat the symptoms of various porphyrias. In addition, it is used in the preparation of agar medium for the bacteria culture.

Solubility
Soluble in dilute ammonia, trimethylamine, p-toluidine and dimethylaniline. Slightly soluble in alcohol. Insoluble in water

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16009-13-5
Molecular Formula C34H32FeN4O4
Molecular Weight (g/mol) 616.50
MDL Number MFCD00010726
InChI Key GGIDWJQWCUJYRY-UHFFFAOYSA-L
Synonym chlorohemin, hemin chloride, protoferriheme, ferriheme, ferriprotoporphyrin ix chloride, ferriprotoporphyrin ix, panhematin, protohemin, protohemin ix, ferriprotoporphyrinShow MoreShow Less
PubChem CID 131675604
SMILES [Fe+3].CC1=C2[N-]C(C=C3N=C(C=C4[N-]C(=CC5=NC(=C2)C(C)=C5C=C)C(C)=C4C=C)C(C)=C3CCC(O)=O)=C1CCC(O)=O

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 5229914
Merck Index 14,4644
Solubility Information Soluble in dilute ammonia,trimethylamine,p-toluidine and dimethylaniline. Slightly soluble in alcohol. Insoluble in water
IUPAC Name 3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethyl-23H-porphyrin-21-id-2-yl]propanoic acid;iron;hydrochloride
Formula Weight 651.95
Percent Purity ≥98%
Chemical Name or Material Hemin (porcine)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253457 Certificate TypeCertificate of Analysis Date19/12/2024 Catalog NumberA11165.03,A11165.14,A11165.06
Lot NumberU21K013 Certificate TypeCertificate of Analysis Date02/10/2024 Catalog NumberA11165.03,A11165.14,A11165.06
Lot Number10251074 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA11165.03,A11165.14,A11165.06
Lot NumberS14K014 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog NumberA11165.03,A11165.14,A11165.06
Lot NumberQ11K010 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberA11165.03,A11165.14,A11165.06
compliance-icons
Product Identifier
  • Hemin (porcine)
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Hemin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hemin (porcine), 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.