missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ Hemispheric Bowl for 100 mL Flask

For Use With: AREX-6 Hotplate Stirrers

Brand:  Velp Scientifica™ A00000331

Product Code. 15895201

  • 164.00 € / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For Use With: AREX-6 Hotplate Stirrers

  • Holds various sizes of flasks
  • For use with 135 mm plate hotplates
Specifications

Specifications

100 mL
Fits on 135 mm plate
AREX-6 Hotplate Stirrers
Bowl
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers