missing translation for 'onlineSavingsMsg'
Learn More

VELP Scientifica™ Hemispheric Bowls for Round Bottom Flasks

The Hemispheric Bowl is a suitable solution for replacing oil baths and mantles in order to perform clean and safe analysis with round bottom flasks.

Brand:  VELP Scientifica™ A00000373

 View more versions of this product

Product Code. 16320528

  • 147.00 € / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Precisely fits 135 mm diameter plate
  • Accommodates different sizes of round bottom flasks
  • Minimizes risk of movement
  • Ensures fastest heat transfer
Specifications

Specifications

Round Bottom Flasks
25 mL
Aluminum
Flask Bowl
Aluminum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers