missing translation for 'onlineSavingsMsg'
Learn More

Heparin Sodium Salt, Thermo Scientific Chemicals

86.18 € - 242.74 €

Chemical Identifiers

CAS 9041-08-1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15815558
View Documents Promotion Details
Thermo Scientific
J16920.B
1 mU Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15885548
View Documents Promotion Details
Thermo Scientific
J16920.!P
100 kU Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15895548
View Documents Promotion Details
Thermo Scientific
J16920.!T
500 kU Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

9041-08-1

Specifications

9041-08-1
Powder
MFCD00081689
Heparin Sodium Salt
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : MI0850000

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only