missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heparin sodium salt, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
Minimum activity: > 150 I.U./mg. Glycosaminoglycan that behaves as an anticoagulant. Binds with high affinity to antithrombin III (AT-III).
Specifications
Specifications
CAS | 9041-08-1 |
MDL Number | MFCD00081689 |
Quantity | 100 mg |
Percent Purity | >98% |
Chemical Name or Material | Heparin sodium salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'