missing translation for 'onlineSavingsMsg'
Learn More

HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™

Commonly used buffering agent

Brand:  Fisher BioReagents BP299-100

38.65 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10041703

  • 44.95 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Commonly used buffering agent

HEPES is commonly used as a buffering agent.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

≤0.01(0.1M solution) at 260nm, 0.004 max. (0.1M solution) at 280nm
1M Solution, pH 7.3
Undesignated
7.2 to 7.5
100 mL
DNase-, RNase- and Protease-Free
C8H18N2O4S
Protease free
N-(2-Hydroxyethyl)piperazine-N«-2-ethanesulfonic Acid
Molecular Biology
HEPES Buffer
7365-45-9, 7732-18-5
0.95 to 1.05 M
Liquid
DNase free
Pass Test
Poly Bottle
15, 4689
238.197
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HEPES Buffer, 1M Solution, pH 7.3

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™ > Poly Bottle; 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.