missing translation for 'onlineSavingsMsg'
Learn More

HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™

Product Code. p-26382
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Glass Bottle
Poly Bottle
Poly Pail
Unit Size:
100g
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10397023

Brand: Fisher BioReagents BP310100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Commonly used buffering agent | CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

Commonly used buffering agent

HEPES is commonly used as a buffering agent.
TRUSTED_SUSTAINABILITY

Specifications

Absorbance 0.01 max. (0.1M solution) at 280nm
Chemical Name or Material HEPES
CAS 7365-45-9
Color White
pH 5.0 to 6.5
Physical Form Powder or Solid
Quantity 100 g
DNase DNase free
Purity Grade Notes DNase-, RNase- and Protease-Free
Assay Percent Range ≥99 %
Identification Pass Test
Molecular Formula C8H18N2O4S
MDL Number MFCD00006158
Packaging Glass Bottle
Protease Protease free
Merck Index 15, 4689
Synonym N-(2-Hydroxyethyl)piperazine-N'-2-ethanesulfonic Acid
Solubility Information Soluble in water
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular Weight (g/mol) 238.30
PubChem CID 23831
ChEBI CHEBI:42334
Percent Purity ≥99%
Grade Molecular Biology
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.