missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals HEPES free acid, 99+%, for molecular biology, DNase, Rnase and Protease free
Click to view available options
Quantity:
50 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Specifications
Absorbance | (0.5 M in water), (0.5 M in water) at 0nm, 0.1 max. at 260nm, 0.1 max. at 280nm |
Absorbance Conditions | (0.5M in water) |
Melting Point | 234.0°C to 238.0°C |
Color | White |
pH | 5 to 6.5 (0.5 M aq. soln.) |
Physical Form | Crystalline Powder |
Quantity | 500 g |
Assay Percent Range | 99+% |
Infrared Spectrum | Authentic |
Loss on Drying | 0.5% max. (105°C) |
Show More |
Safety and Handling
Product Identifier
- HEPES
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction