missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES free acid, 99+%, for molecular biology, DNase, Rnase and Protease free

Product Code. 10723894
Click to view available options
Quantity:
50 g
500 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Absorbance (0.5 M in water), (0.5 M in water) at 0nm, 0.1 max. at 260nm, 0.1 max. at 280nm
Absorbance Conditions (0.5M in water)
Melting Point 234.0°C to 238.0°C
Color White
pH 5 to 6.5 (0.5 M aq. soln.)
Physical Form Crystalline Powder
Quantity 50 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (105°C)
Packaging Plastic bottle
Beilstein 23, V,2, 379
Merck Index 15, 4689
Solubility Information Solubility in water: 704g/L (20°C).
Formula Weight 238.3
Percent Purity ≥99%
Grade Molecular Biology
Chemical Name or Material HEPES free acid
Show More Show Less
Product Identifier
  • HEPES

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.