missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HEPES Lysis Buffer with NP-40

40.20 € - 72.15 €

Specifications

Content And Storage Ambient temperatures
Format Liquid
Product Type Cell Lysis Reagent
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15465209
View Documents
Thermo Scientific™
J62915.AP
500mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15455209
View Documents
Thermo Scientific™
J62915.AK
250mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ambient temperatures
Liquid
Cell Lysis Reagent
Videos
SDS
Documents

Documents

Product Certifications