missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES sodium salt, 99%

Product Code. 11447837
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

HEPES [N-(2-hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid)] is a general purpose buffer for biological research. Its applications include, tissue culture, Phosphorylation and photophosphorylation, fixative Transmission electron microscopy, protein synthesis, preventing binding to non-receptor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES [N-(2-hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)] is a general purpose buffer for biological research. Its applications include, tissue culture, Phosphorylation and photophosphorylation, fixative Transmission electron microscopy, protein synthesis, preventing binding to non-receptor materials.

Solubility
It is soluble in water.

Notes
Stable under ordinary conditions. Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym hepes sodium salt, hepes hemisodium salt, sodium 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonate, unii-z9fto91o8a, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl-, monosodium salt, z9fto91o8a, sodium 4-2-hydroxyethyl piperazin-1-ylethanesulphonate, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid sodium salt, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid hemisodium salt
PubChem CID 2724248
ChEBI CHEBI:46758
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Quantity 25 g
Sensitivity Hygroscopic
Solubility Information It is soluble in water.
IUPAC Name sodium;2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonate
Formula Weight 260.28
Percent Purity 99%
Chemical Name or Material HEPES sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'