missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals HEPES sodium salt, 99% Product Code.: 11457837

Thermo Scientific Chemicals HEPES sodium salt, 99%

Product Code. 11457837
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11457837

Brand: Thermo Scientific Chemicals A16516.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HEPES [N-(2-hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid)] is a general purpose buffer for biological research. Its applications include, tissue culture, Phosphorylation and photophosphorylation, fixative Transmission electron microscopy, protein synthesis, preventing binding to non-receptor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES [N-(2-hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)] is a general purpose buffer for biological research. Its applications include, tissue culture, Phosphorylation and photophosphorylation, fixative Transmission electron microscopy, protein synthesis, preventing binding to non-receptor materials.

Solubility
It is soluble in water.

Notes
Stable under ordinary conditions. Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym hepes sodium salt, hepes hemisodium salt, sodium 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonate, unii-z9fto91o8a, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl-, monosodium salt, z9fto91o8a, sodium 4-2-hydroxyethyl piperazin-1-ylethanesulphonate, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid sodium salt, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid hemisodium salt
PubChem CID 2724248
ChEBI CHEBI:46758
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Quantity 100 g
Sensitivity Hygroscopic
Solubility Information It is soluble in water.
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
Formula Weight 260.28
Percent Purity 99%
Chemical Name or Material HEPES sodium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0455462 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog NumberA16516.14,A16516.22
Lot NumberA0447794 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog NumberA16516.14,A16516.22
Lot Number10243600 Certificate TypeCertificate of Analysis Date21/09/2022 Catalog NumberA16516.14,A16516.22
Lot Number10237910 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA16516.14,A16516.22
Lot Number10186651 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16516.14,A16516.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals HEPES sodium salt, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.