missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals HEPES sodium salt, 99%
51.15 € - 885.41 €
Chemical Identifiers
CAS | 75277-39-3 |
---|---|
Molecular Formula | C8H17N2NaO4S |
Molecular Weight (g/mol) | 260.28 |
MDL Number | MFCD00036463 |
InChI Key | RDZTWEVXRGYCFV-UHFFFAOYSA-M |
Synonym | 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid sodium salt |
PubChem CID | 2724248 |
ChEBI | CHEBI:46758 |
IUPAC Name | sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate |
SMILES | [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10695852
|
Thermo Scientific Chemicals
215000250 |
25 g |
51.15 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10411451
|
Thermo Scientific Chemicals
215001000 |
100 g |
120.60 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10542714
|
Thermo Scientific Chemicals
215005000 |
500 g |
477.38 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10532554
|
Thermo Scientific Chemicals
215000010 |
1 kg |
885.41 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
75277-39-3 | |
260.28 | |
RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
2724248 | |
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate |
C8H17N2NaO4S | |
MFCD00036463 | |
2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid sodium salt | |
CHEBI:46758 | |
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 |
Specifications
(0.1 M aq. soln.), (0.1 M aq. soln.) at 0nm, 0.02 at 280nm, 0.04 at 260nm | |
75277-39-3 | |
9.5 to 10.5 (1% aq. soln.) | |
99% | |
C8H17N2NaO4S | |
Glass bottle | |
RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate | |
2724248 | |
260.27 | |
HEPES sodium salt, 99% |
(0.1M aq. soln.) | |
White | |
Crystalline Powder | |
Authentic | |
MFCD00036463 | |
2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid sodium salt | |
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 | |
260.28 | |
CHEBI:46758 | |
≥98.5% (on anhydrous substance) |
Safety and Handling
missing translation for 'einecsNumber' : 278-169-7
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title