missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free

Product Code. 10328103
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid sodium salt
PubChem CID 2724248
ChEBI CHEBI:46758
IUPAC Name sodium;2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Absorbance (0.1 M in H2O), (0.1 M in H2O) at 0nm, 0.02 max. at 280nm, 0.04 max. at 260nm
Absorbance Conditions (0.1M in H2O)
Melting Point 248.0°C
Color White
pH 5.0 to 15.0 (1% soln.)
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Packaging Plastic bottle
Formula Weight 260.27
Percent Purity ≥99%
Grade Molecular Biology
Chemical Name or Material HEPES sodium salt, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.