missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyric acid, 99%, for protein sequence analysis, Thermo Scientific Chemicals

Product Code. 10674012
Click to view available options
Quantity:
100 mL
150 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-22-4
Molecular Formula C4HF7O2
Molecular Weight (g/mol) 214.04
MDL Number MFCD00004171
InChI Key YPJUNDFVDDCYIH-UHFFFAOYSA-N
Synonym heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
PubChem CID 9777
ChEBI CHEBI:39426
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoic acid
SMILES C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O

Specifications

Absorbance (0.1% aqueous solution), (0.1% aqueous solution) at 0nm, 0.004 max. at 280nm, 0.006 max. at 254nm, 0.15 max. at 230nm
Melting Point -17.5°C
Color Beige to Brown-Orange
Density 1.6400g/mL
Boiling Point 120.0°C (755.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CF3CF2CF2CO2H
Refractive Index 1.3
Quantity 150 mL
Specific Gravity 1.64
Solubility Information Solubility in water: miscible. Other solubilities: soluble in benzene and carbon tetrachloride, miscible with acetone, ether and petroleum ether, insoluble in carbon disulfide and mineral oil
Formula Weight 214.04
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Heptafluorobutyric acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'