missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane, 99%, Reagent Grade, Honeywell™
Reagent Grade, 99%
44.39 € - 5703.53 €
Chemical Identifiers
CAS | 142-82-5 |
---|---|
Molecular Formula | C7H16 |
Molecular Weight (g/mol) | 100.21 |
MDL Number | MFCD00009544 |
InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
Synonym | n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish |
PubChem CID | 8900 |
ChEBI | CHEBI:43098 |
IUPAC Name | heptane |
SMILES | CCCCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15667900
|
Honeywell Research Chemicals
H2198-500ML |
500 mL | Glass Bottle | ||||||
15607900
|
Honeywell Research Chemicals
H2198-1L |
1 L | Glass Bottle | ||||||
15617900
|
Honeywell Research Chemicals
H2198-2.5L |
2.5 L | Glass Bottle |
223.94 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15657900
|
Honeywell Research Chemicals
H2198-4X2.5L |
4 x 2.5 L | Glass Bottle | ||||||
15647900
|
Honeywell Research Chemicals
H2198-20L |
20 L | Stainless Steel Can | ||||||
15627900
|
Honeywell Research Chemicals
H2198-200L |
200 L | Metal Drum | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
142-82-5 | |
100.21 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
8900 | |
heptane |
C7H16 | |
MFCD00009544 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CHEBI:43098 | |
CCCCCCC |
Specifications
-90.5°C | |
Colorless | |
CH3(CH2)5CH3 | |
UN1206 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
heptane | |
8900 | |
100.2g/mol | |
0.684g/mL at 25°C | |
0.07 | |
433°F | |
Liquid | |
Heptane |
142-82-5 | |
C7H16 | |
MFCD00009544 | |
1730763 | |
0.003g/L in water at 0°C | |
CCCCCCC | |
100.21 | |
CHEBI:43098 | |
0.99 | |
98.2°C to 98.4°C (1000 hPa) | |
−4°C (24.8°F) | |
Characteristic | |
83mmHg (37.7°C), 40mmHg (20°C) |
Safety and Handling
P210-P301 + P310-P331-P370 + P378-P403 + P235
missing translation for 'dotInformation' : class 3 - PG 2 - Heptanes
missing translation for 'rtecsNumber' : 205-563-8