missing translation for 'onlineSavingsMsg'
Learn More

Heptane Fraction, Honeywell Riedel-de Haën™

Product Code. 15685730
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Aluminum Bottle
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

boiling range 93.5 - 100.4 °C

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-82-5
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098

Specifications

Melting Point -90.5°C
Molecular Formula C7H16
MDL Number MFCD00009544
UN Number UN1206
Beilstein 1730763
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES CCCCCCC
IUPAC Name heptane
Molecular Weight (g/mol) 100.21
Density 0.71g/mL at 20°C
Boiling Point 98.2°C to 98.4°C
Quantity 1 L
Flash Point −4°C (24.8°F)
Packaging Glass Bottle
Refractive Index 1.397
Chemical Name or Material Heptane Fraction
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'