missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane-(n) 99+%, HPLC Grade For HPLC and Laboratory Use, Chem Lab
Click to view available options
Quantity:
1 L
2.5 L
Specifications
Specifications
Melting Point | -90°C |
Color | Colorless |
Acidity | < 0.0005 meq/g |
Density | 0.69 g/mol |
Boiling Point | 98°C |
Quantity | 2.5 L |
Flash Point | -4 °C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Assay Percent Range | > 99% |
Show More |
Safety and Handling




Product Identifier
- Heptane-(n) 99+%, HPLC grade
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P331-Do NOT induce vomiting.
- P403+P235-Store in a well-ventilated place. Keep cool.
- MIXTURE LIST-Contains : Heptane-(n)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Heptane-(n) 99+%, HPLC Grade For HPLC and Laboratory Use, Chem Lab >
Spot an opportunity for improvement?Share a Content Correction