Learn More
HER2/ErbB2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 51105-1-AP-20UL
 View more versions of this product
Product Code. 16836395
  • 145.00 € / 20µL
Explore more special offers
Description
Description
This protein is a regulatory subunit of the condensin-2 complex, a complex which establishes mitotic chromosome architecture and is involved in physical rigidity of the chromatid axis.
Specifications
Specifications
HER2/ErbB2
Polyclonal
Unconjugated
ERBB2
CD340, ERBB2, HER 2, HER 2/neu, HER2, MLN 19, MLN19, NEU, NGL, p185, p185erbB2, Proto oncogene c ErbB 2, Proto oncogene Neu, TKR1
Rabbit
Antigen Affinity Chromatography
RUO
2064
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P04626
ERBB2
Peptide
20 μL