missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Heratherm Stability Chamber 750 L
Gain confidence in your product when testing stability and shelf life with robust Heratherm Stability chambers designed to meet a variety of quality control demands.
Specifications
Alarm Types | Audible and Visible |
---|---|
Capacity (English) Shelf | 66.1 lbs |
Capacity (Metric) Shelf | 30 kg |
Certifications/Compliance | UL, CE |
Data Outputs | USB, Ethernet |
Product Code | Brand | Door Style | Shipping Weight (English) | Shipping Weight (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Door Style | Shipping Weight (English) | Shipping Weight (Metric) | Price | Quantity & Availability | |||
18724594
|
Thermo Scientific™
ESS750 |
Solid outer door with inner glass door | 840 lbs | 381 kg |
N/A
|
N/A | |||
18704594
|
Thermo Scientific™
ESG750 |
Glass door | 820 lbs | 372 kg |
N/A
|
N/A | |||
Description
Temperature and humidity controls, well suited for stability testing, shelf life testing, and environmental testing.
- Broad relative humidity range to cover many applications, including humidity levels below ambient.
- Environmentally friendly water-efficient humidification system.
- Minimal condensate in standard applications.
- Integrated water tank; fixed water connection for in-house purified water option available.
- Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
- Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
- Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard.
- Sample and process protection: Assign personalized access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
- Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
- Simplified setup with no technician required; install using standard power cord.
- IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
- Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
- Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
- Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)
Specifications
Audible and Visible | |
30 kg | |
USB, Ethernet | |
Powder-coated Cold Rolled Steel | |
3 | |
10 to 90% | |
Humidity Chamber | |
10 - 90% RH | |
2 access ports | |
0.46 m² | |
Microprocessor - programmable | |
≤ ±0.1°C @ 20°C to 37°C | |
24 months | |
0.05 - 1 MΩ | |
33.07 in. | |
1080 mm | |
795 mm | |
57.09 in | |
1450 mm | |
24.8 in. | |
630 mm | |
100/115/220/230 V |
66.1 lbs | |
UL, CE | |
750 L | |
Powder-coated Cold Rolled Steel | |
Nema 5-20P | |
0 to 70°C | |
7 in. touchscreen user interface | |
4 | |
Temperature and humidity ramping | |
Stainless Steel (304/1.4301) | |
0.1°C | |
≤ ±0.5°C @ 20°C to 37°C | |
0.15 to 1 L per day | |
42.52 in. | |
22.6 in. | |
840 mm | |
72.24 in. | |
1835 mm | |
34.84 in. | |
885 mm | |
100 - 240V, 50-60Hz, 1400W | |
1400 W |