missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Heratherm Stacking Kit

Stacking kit for 2 100L or 60L on top of 100L Heratherm ovens and incubators

Brand:  Thermo Scientific™ 50126666

Product Code. 10801311

  • 341.70 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Stacking kit for 2 100L or 60L on top of 100L Heratherm ovens and incubators. Recommended if performing decontaminiantion cycle in lower unit or stacking an incubator on an oven

Heratherm Stacking kit

For 2 100L or 60L on top of 100L Heratherm ovens and incubators
Recommended if performing decontaminiantion cycle in lower unit or stacking an incubator on an oven

Suitable for use with:
General Protocol
Advanced Protocol
Advanced Protocol Security ovens

448 x 511 mm

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 L or 60 L on top of 100 L Heratherm ovens and incubators
100 L or 60 L
Oven Accessories
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.