missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Herbimycin A
Brand: Novus Biologicals™ NB110-81565
This item is not returnable.
View return policy
Specifications
Store at -20 C. Avoid freeze-thaw cycles. | |
Soluble in DMSO and Ethanol (1 mg/mL). | |
Herbimycin |
0.1 mg | |
Herbimycin A |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction