missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Hesperidin, 97%, includes analogeous compounds

Product Code. 10376520 Shop All Thermo Scientific Temperature Control Solutions Products
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 520-26-3
Molecular Formula C28H34O15
Molecular Weight (g/mol) 610.57
MDL Number MFCD00075663
InChI Key QUQPHWDTPGMPEX-QJBIFVCTSA-N
PubChem CID 45358131
SMILES COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1

Specifications

Melting Point 258.0°C to 262.0°C
Color Brown to White
Loss on Drying 4% max.
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 5 g
Merck Index 15, 4706
Specific Rotation − 76.00
Solubility Information Solubility in water: 1g/50l. Other solubilities: soluble in diluted alkalies,soluble in pyridine,soluble in formamide and dimethylformamide (60 c),slightly sol. in methanol and glacial acetic acid,practically insol. in acetone benzene and chcl3
IUPAC Name (2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-3,4-dihydro-2H-1-benzopyran-4-one
Formula Weight 610.54
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Hesperidin, 97%
Show More Show Less
Product Identifier
  • Hesperidin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.