missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Hesperidin, 97%, includes analogeous compounds
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 258.0°C to 262.0°C |
Color | Brown to White |
Loss on Drying | 4% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Plastic bottle |
Quantity | 250 g |
Merck Index | 15, 4706 |
Specific Rotation | − 76.00 |
Solubility Information | Solubility in water: 1g/50l. Other solubilities: soluble in diluted alkalies,soluble in pyridine,soluble in formamide and dimethylformamide (60 c),slightly sol. in methanol and glacial acetic acid,practically insol. in acetone benzene and chcl3 |
Show More |
Safety and Handling
Product Identifier
- Hesperidin
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'