missing translation for 'onlineSavingsMsg'
Learn More

Hexa-n-butylditin, 97%, Thermo Scientific Chemicals

51.26 € - 614.06 €

Chemical Identifiers

CAS 813-19-4
Molecular Formula C24H54Sn2
Molecular Weight (g/mol) 580.12
MDL Number MFCD00009417
InChI Key REDSKZBUUUQMSK-UHFFFAOYSA-N
Synonym hexabutylditin, hexabutyldistannane, bis, distannane, hexabutyl, tin, hexabutyldi, stannane, hexabutyldi, hexa-n-butylditin, 1,1,1,2,2,2-hexabutyldistannane, hexabutyidistannane, hexabutyl distannane
PubChem CID 6327815
IUPAC Name tributyltin
SMILES CCCC[Sn](CCCC)(CCCC)[Sn](CCCC)(CCCC)CCCC
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11494054
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12007.09
10 g
51.26 €
10g
Please to purchase this item. Need a web account? Register with us today!
11411900
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12007.14
25 g
103.52 €
25g
Please to purchase this item. Need a web account? Register with us today!
11404064
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12007.18
50 g
174.87 €
50g
Please to purchase this item. Need a web account? Register with us today!
11414064
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12007.30
250 g
614.06 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

813-19-4
580.12
REDSKZBUUUQMSK-UHFFFAOYSA-N
6327815
CCCC[Sn](CCCC)(CCCC)[Sn](CCCC)(CCCC)CCCC
C24H54Sn2
MFCD00009417
hexabutylditin, hexabutyldistannane, bis, distannane, hexabutyl, tin, hexabutyldi, stannane, hexabutyldi, hexa-n-butylditin, 1,1,1,2,2,2-hexabutyldistannane, hexabutyidistannane, hexabutyl distannane
tributyltin

Specifications

813-19-4
197°C to 198°C (10 mmHg)
C24H54Sn2
1.512
UN2788
Air and moisture sensitive
Immiscible with water.
CCCC[Sn](CCCC)(CCCC)[Sn](CCCC)(CCCC)CCCC
580.12
580.07
Hexa-n-butylditin
1.152
124°C (255°F)
(CH3CH2CH2CH2)3Sn-Sn(CH2CH2CH2CH3)3
MFCD00009417
3605476
hexabutylditin, hexabutyldistannane, bis, distannane, hexabutyl, tin, hexabutyldi, stannane, hexabutyldi, hexa-n-butylditin, 1,1,1,2,2,2-hexabutyldistannane, hexabutyidistannane, hexabutyl distannane
REDSKZBUUUQMSK-UHFFFAOYSA-N
tributyltin
6327815
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P501c

H301-H311-H315-H319-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

missing translation for 'einecsNumber' : 212-383-3

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only