missing translation for 'onlineSavingsMsg'
Learn More
Hexadecyltrimethylammonium bromide, 99+%, Thermo Scientific Chemicals Product Code.: 10541671

Hexadecyltrimethylammonium bromide, 99+%, Thermo Scientific Chemicals

Product Code. 10541671
500 g, Plastic Bottle
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10541671

Brand: Thermo Scientific Acros 227165000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-09-0
Molecular Formula C19H42BrN
Molecular Weight (g/mol) 364.46
MDL Number MFCD00011772
InChI Key LZZYPRNAOMGNLH-UHFFFAOYSA-M
PubChem CID 5974
ChEBI CHEBI:3567
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Color White
Physical Form Crystalline Powder
Melting Point 230.0°C
Decomposition Information >230°C
Quantity 500 g
Flash Point 244°C
Infrared Spectrum Authentic
Packaging Plastic Bottle
Assay Percent Range 99+%
Linear Formula CH3(CH2)15N(CH3)3Br
Beilstein 04, IV, 819
Fieser 15,77
Merck Index 15, 2024
Synonym Cetrimonium Bromide, Cetyltrimethylammonium Bromide, CTABr
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: soluble in methanol (>200g/L at 20 °C), ethanol, insoluble in benzene, diethyl ether, chloroform, sparingly soluble in acetone
IUPAC Name hexadecyl(trimethyl)azanium;bromide
Formula Weight 364.45
Percent Purity ≥99%
Chemical Name or Material Hexadecyltrimethylammonium Bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexadecyltrimethylammonium bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hexadecyltrimethylammonium bromide, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.