missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexafluoro-2-(p-tolyl)-2-propanol, 97%, Thermo Scientific Chemicals
72.70 € - 281.00 €
Chemical Identifiers
CAS | 2010-61-9 |
---|---|
Molecular Formula | C10H8F6O |
Molecular Weight (g/mol) | 258.16 |
MDL Number | MFCD00015357 |
InChI Key | AOAVZPXKNQAALI-UHFFFAOYSA-N |
Synonym | hexafluoro-2-p-tolyl-2-propanol, 2-p-tolyl hexafluoroisopropanol, hexafluoro-2-4-tolyl isopropanol, hexafluoro-2-p-tolyl isopropanol, 1,1,1,3,3,3-hexafluoro-2-4-methylphenyl propan-2-ol, benzenemethanol, 4-methyl-.alpha.,.alpha.-bis trifluoromethyl, 4-methyl-alpha,alpha-bis trifluoromethyl benzyl alcohol, hexafluoro-2 p-tolyl isopropanol, benzenemethanol, 4-methyl-alpha,alpha-bis trifluoromethyl, hexafluoro-2-p-tolyl-i-propanol |
PubChem CID | 74829 |
IUPAC Name | 1,1,1,3,3,3-hexafluoro-2-(4-methylphenyl)propan-2-ol |
SMILES | CC1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2010-61-9 | |
258.16 | |
AOAVZPXKNQAALI-UHFFFAOYSA-N | |
74829 | |
CC1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F |
C10H8F6O | |
MFCD00015357 | |
hexafluoro-2-p-tolyl-2-propanol, 2-p-tolyl hexafluoroisopropanol, hexafluoro-2-4-tolyl isopropanol, hexafluoro-2-p-tolyl isopropanol, 1,1,1,3,3,3-hexafluoro-2-4-methylphenyl propan-2-ol, benzenemethanol, 4-methyl-.alpha.,.alpha.-bis trifluoromethyl, 4-methyl-alpha,alpha-bis trifluoromethyl benzyl alcohol, hexafluoro-2 p-tolyl isopropanol, benzenemethanol, 4-methyl-alpha,alpha-bis trifluoromethyl, hexafluoro-2-p-tolyl-i-propanol | |
1,1,1,3,3,3-hexafluoro-2-(4-methylphenyl)propan-2-ol |
Specifications
2010-61-9 | |
C10H8F6O | |
MFCD00015357 | |
hexafluoro-2-p-tolyl-2-propanol, 2-p-tolyl hexafluoroisopropanol, hexafluoro-2-4-tolyl isopropanol, hexafluoro-2-p-tolyl isopropanol, 1,1,1,3,3,3-hexafluoro-2-4-methylphenyl propan-2-ol, benzenemethanol, 4-methyl-.alpha.,.alpha.-bis trifluoromethyl, 4-methyl-alpha,alpha-bis trifluoromethyl benzyl alcohol, hexafluoro-2 p-tolyl isopropanol, benzenemethanol, 4-methyl-alpha,alpha-bis trifluoromethyl, hexafluoro-2-p-tolyl-i-propanol | |
CC1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F | |
258.16 | |
258.17 | |
Hexafluoro-2-(p-tolyl)-2-propanol |
181°C to 182°C | |
1.424 | |
2127760 | |
AOAVZPXKNQAALI-UHFFFAOYSA-N | |
1,1,1,3,3,3-hexafluoro-2-(4-methylphenyl)propan-2-ol | |
74829 | |
97% |
Safety and Handling
missing translation for 'einecsNumber' : 217-928-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only