missing translation for 'onlineSavingsMsg'
Learn More

Hexafluorobenzene, 99%, Thermo Scientific Chemicals

Product Code. 10209503
Click to view available options
Quantity:
10 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392-56-3
Molecular Formula C6F6
Molecular Weight (g/mol) 186.06
MDL Number MFCD00000288
InChI Key ZQBFAOFFOQMSGJ-UHFFFAOYSA-N
Synonym hexafluorobenzene, perfluorobenzene, benzene, hexafluoro, hexafluorbenzol, unii-cmc18t611k, benzene, 1,2,3,4,5,6-hexafluoro, hexa fluorobenzene, hexafluoro benzene, pubchem18879, acmc-1bmus
PubChem CID 9805
ChEBI CHEBI:38589
IUPAC Name 1,2,3,4,5,6-hexafluorobenzene
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)F

Specifications

Melting Point 3.7°C to 4.1°C
Color Colorless to Yellow
Density 1.6120g/mL
Boiling Point 81.0°C to 82.0°C (743.0 mmHg)
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6F6
Refractive Index 1.3760 to 1.3780
Quantity 100 mL
Merck Index 15,4722
Specific Gravity 1.612
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in ethanol
Formula Weight 186.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Hexafluorobenzene
Show More Show Less
compliance-icons
Product Identifier
  • Hexafluorobenzene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'