missing translation for 'onlineSavingsMsg'
Learn More
Hexahydro-4-methylphthalic anhydride, 98%, mixture of cis and trans, Thermo Scientific Chemicals Product Code.: 10605992

Hexahydro-4-methylphthalic anhydride, 98%, mixture of cis and trans, Thermo Scientific Chemicals

Product Code. 10605992
500 g, Glass bottle
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10605992

Brand: Thermo Scientific Acros 165395000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19438-60-9
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00005927
InChI Key FKBMTBAXDISZGN-UHFFFAOYSA-N
Synonym hexahydro-4-methylphthalic anhydride, 4-methylhexahydrophthalic anhydride, 5-methylhexahydroisobenzofuran-1,3-dione, 4-methyl-1,2-cyclohexanedicarboxylic anhydride, 1,3-isobenzofurandione, hexahydro-5-methyl, 1,3-isobenzofurandione, hexahydromethyl, 4-methylcyclohexane-1,2-dicarboxylic anhydride, 5-methylhexahydrophthalic anhydride, 5-methyl-3a,4,5,6,7,7a-hexahydroisobenzofuran-1,3-dione, 4-methyl-1,2-cyclohexanedicarboxylic anhydride, mixture of isomersShow MoreShow Less
PubChem CID 86876
IUPAC Name 5-methyl-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES CC1CCC2C(C1)C(=O)OC2=O

Specifications

Melting Point -29°C
Density 1.1600g/mL
Boiling Point 120°C (1.0 Mbar)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4764 to 1.4784
Quantity 500 g
Specific Gravity 1.16
Solubility Information Solubility in water: reacts with water
Viscosity 65 mPa.s (20°C)
Formula Weight 168.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Hexahydro-4-methylphthalic anhydride, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0450668 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number165395000,165390025
Lot NumberA0401246 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number165395000,165390025
Lot NumberA0400500 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number165395000,165390025
Lot NumberA0447202 Certificate TypeCertificate of Analysis Date16/11/2022 Catalog Number165395000,165390025
Lot NumberA0435383 Certificate TypeCertificate of Analysis Date18/10/2021 Catalog Number165395000,165390025
compliance-icons compliance-icons
Product Identifier
  • Hexahydro-4-methylphthalic anhydride, mixture of cis and trans
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hexahydro-4-methylphthalic anhydride, 98%, mixture of cis and trans, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.