missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyldisiloxane, 98+%, Thermo Scientific Chemicals

Product Code. 10523371
Click to view available options
Quantity:
10 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-46-0
InChI Key UQEAIHBTYFGYIE-UHFFFAOYSA-N
Synonym hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide
PubChem CID 24764
ChEBI CHEBI:78002
IUPAC Name trimethyl(trimethylsilyloxy)silane
SMILES C[Si](C)(C)O[Si](C)(C)C

Specifications

Color Colorless
Density 0.7600g/mL
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C6H18OSi2
Linear Formula (CH3)3SiOSi(CH3)3
Refractive Index 1.3765 to 1.3785 (20°C, 589nm)
Quantity 10 mL
Fieser 08,240; 09,55; 10,196
Molecular Weight (g/mol) 162.379
Formula Weight 162.38
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexamethyldisiloxane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.