missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyleneimine, 99%

Product Code. 10741735 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
500 mL
Packaging:
Glass bottle
Unit Size:
25mL
500mL
5mL
This item is not returnable. View return policy

Product Code. 10741735

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-49-9 | C6H13N | 99.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-49-9
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.17
MDL Number MFCD00006934
InChI Key ZSIQJIWKELUFRJ-UHFFFAOYSA-N
Synonym hexamethyleneimine, homopiperidine, hexahydroazepine, hexahydro-1h-azepine, perhydroazepine, 1h-azepine, hexahydro, hexamethylenimine, azacycloheptane, azepan, hexamethylene imine
PubChem CID 8119
ChEBI CHEBI:32616
IUPAC Name azepane
SMILES C1CCCNCC1

Specifications

Melting Point -37.0°C
Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 138.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4650 to 1.4670
Quantity 25 mL
Beilstein 20, 94
Specific Gravity 0.88
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in strongly alkaline materials, soluble in methanol, ethanol, benzene and ether
Formula Weight 99.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Hexamethyleneimine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexamethyleneimine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.