missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamine, Certified AR for Analysis, Fisher Chemical™
73.55 € - 313.00 €
Chemical Identifiers
CAS | 100-97-0 |
---|---|
Molecular Formula | C6H12N4 |
Molecular Weight (g/mol) | 140.19 |
MDL Number | 6895 |
InChI Key | VKYKSIONXSXAKP-UHFFFAOYSA-N |
Synonym | methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine |
PubChem CID | 4101 |
ChEBI | CHEBI:6824 |
SMILES | C1N2CN3CN1CN(C2)C3 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10755331
|
Fisher Chemical
H/0320/53 |
500 g | Plastic powder jar |
73.55 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10315900
|
Fisher Chemical
H/0320/60 |
1 kg | Plastic powder jar |
93.40 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10645682
|
Fisher Chemical
H/0320/65 |
5 kg | Plastic keg |
313.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Hexamine, Certified AR for Analysis
Chemical Identifiers
100-97-0 | |
140.19 | |
VKYKSIONXSXAKP-UHFFFAOYSA-N | |
4101 | |
C1N2CN3CN1CN(C2)C3 |
C6H12N4 | |
6895 | |
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine | |
CHEBI:6824 |
Specifications
100-97-0 | |
8.5 to 9.5 | |
6895 | |
VKYKSIONXSXAKP-UHFFFAOYSA-N | |
140.19 | |
CHEBI:6824 | |
Solid |
White | |
C6H12N4 | |
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine | |
C1N2CN3CN1CN(C2)C3 | |
4101 | |
140.19g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Hexamethylenetetramine
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/A§.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title