missing translation for 'onlineSavingsMsg'
Learn More

Hexanal, 96%, stabilized, Thermo Scientific Chemicals

Product Code. 10735821
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-25-1, 59-02-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00007027
InChI Key JARKCYVAAOWBJS-UHFFFAOYSA-N
Synonym caproaldehyde, hexaldehyde, caproic aldehyde, capronaldehyde, 1-hexanal, n-hexanal, hexanaldehyde, n-caproaldehyde, hexylaldehyde, aldehyde c-6
PubChem CID 6184
IUPAC Name hexanal
SMILES CCCCCC=O

Specifications

Acidity 10mg KOH/g max.
Melting Point -56.0°C
Color Colorless to Yellow
Density 0.8150g/mL
Boiling Point 130.0°C to 131.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CHO
Refractive Index 1.4020 to 1.4070
Quantity 1 L
Beilstein 01, II, 745
Merck Index 15, 1762
Specific Gravity 0.815
Solubility Information Solubility in water: 4.8g/L (20°C). Other solubilities: soluble in methanol, acetone, chloroform, ether
Stabilizer approx. 500ppm alpha-tocopherol
Viscosity 0.69 mPa.s (20°C)
Formula Weight 100.16
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Hexanal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hexanal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'