missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, 95% n-Hexane, BAKER ANALYZED, J.T. Baker™
High quality grade chemical for laboratory and specialized industrial use.
79.25 € - 324.00 €
Chemical Identifiers
CAS | 110-54-3 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD02179311 |
InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
PubChem CID | 8058 |
ChEBI | CHEBI:29021 |
IUPAC Name | hexane |
SMILES | CCCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10664631
|
JT Baker
8044.1000 |
1 L | Glass |
79.25 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10507472
|
JT Baker
8044.5000 |
5 L | EcoTainer Metal Safety Can |
324.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Label provides an actual Lot Analysis as a guarantee of purity and consistency
- Analytical methods used for these determinations are based on standard test procedures described in technical publications
- Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability
Chemical Identifiers
110-54-3 | |
86.18 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
8058 | |
hexane |
C6H14 | |
MFCD02179311 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CHEBI:29021 | |
CCCCCC |
Specifications
-95°C | |
Colorless | |
CH3(CH2)4CH3 | |
1208 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
hexane | |
8058 | |
86.18g/mol | |
BAKER ANALYZED™ | |
69°C | |
−26°C | |
Liquid | |
0.326 mPas (dynamic), 0.50mm2/s (kinematic) |
110-54-3 | |
C6H14 | |
MFCD02179311 | |
1730733 | |
0.1g/L in water | |
CCCCCC | |
86.18 | |
CHEBI:29021 | |
≥95% | |
0.657 g/mL at 25°C | |
1.2 - 7.4 Vol % | |
240°C | |
Max. 0.001% | |
n-Hexane |
Safety and Handling
missing translation for 'einecsNumber' : 203-777-6
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.